Sharps Disposal Containers, Large Volume, Covidien
Proveedor: Covidien
8980- 8938 8980S 8997SPG2 8991 8998 8935 8997
58578-090CS 6949.64 MXN
15704-295 89004-914 58578-092 15704-212 58578-090 82003-550 15704-216 15704-194
Sharps Disposal Containers, Large Volume, Covidien
Recipientes Contenedores de objetos punzantes
Red containers offer both large and small openings for infectious and biohazardous waste of various types
Containers meet or exceed JCAHO, OSHA, and NIOSH/CDC standards and requirements.
Containers with hinged lids can easily accommodate the largest sharps suction canisters, trocars, endoscopic instruments, and more. Hinged lids permit temporary closure when not in use. All containers lock for final disposal. Containers 82003-550 and 15704-216 feature a hinged, sealing gasket lid to provide leak-resistant containment of contents. Container 89004-914 features a gasketed, slide-top lid. It is PG II rated and DOT approved for transport of regulated medical waste including cultures and stocks.