VWR® English/Spanish Good Laboratory Practice Notebooks
Proveedor: VWR
VWR®
818-0094 818-0098 818-0096 818-0093 818-0099 818-0095
89005-158EA 934.07 MXN
89005-158 89005-168 89005-164 89005-162 89005-160 89005-170
VWR® English/Spanish Good Laboratory Practice Notebooks
Cuadernos
Bilingual English/Spanish laboratory notebooks feature user data, guidelines, table of contents, and documentation pages with researcher and witness signature blocks.
  • Cover color: Royal blue
  • Tamper-evident section-sewn binding
Acid-free, archival-safe paper pages are formatted in either 5mm (3/16") grid or 5mm (3/16") horizontal rule. Case bound in an imitation leather cover specially coated for durability in laboratory environments.