VWR® Centrifuge Bottles with Caps
Proveedor: Avantor
VWR®
47735-702 47735-688 47735-696 47735-694 47735-692 47735-698 47735-690 47735-700
47735-690CS 11534.88 MXN
47735-690 47735-700 47735-702 47735-696 47735-688 47735-698 47735-692 47735-694
VWR® Centrifuge Bottles with Caps
Frascos Frascos de centrífuga
Centrifuge bottles are manufactured from polycarbonate or polypropylene.
  • Great chemical resistance
  • Caps are fabricated from polypropylene or noryl
Precaución: Maximum RCF ratings are based on using the proper rotor and/or support cushions. Without the proper support, these bottles may distort and/or fail.