VWR® Transparent Plastic Laboratory Aprons
Proveedor: VWR
VWR®
6110 1120 1130 2140 2150 1110
10825-061CS 39258.24 MXN
10824-125 10825-048 10825-061 10824-067 10824-089 10824-045
VWR® Transparent Plastic Laboratory Aprons
Ropa de trabajo
Made of soft, strong 100% polyvinylchloride (PVC) with electronically sealed ties and seams.
  • Protect against water, oil, grease, and most chemicals
  • Will not peel, curl, or crack
  • Soft, pliable Plastipron® design
Ties are double anchored with heat-sealed PVC grommets. Easily cleaned with a damp cloth or sponge.
Información para pedidos: Packed in individual sealed and labeled plastic bags.