VWR® Bottle Tote™ Safety Carriers
Proveedor: Avantor
VWR® Bottle Tote™
B100-1 B-101 B-100 B-102 B102-1
56608-975EA 2043.47 MXN
56608-975 56608-972 56608-970 56608-982 56608-980
VWR® Bottle Tote™ Safety Carriers
Portadores Portadores de frascos
Carriers are designed for the safe transport of acids, alkalies, and solvents.
  • High degree of tensile strength
  • Centering ring assures proper clearance for safe transport
  • Molded-in handle
High in tensile strength, carriers can be used in temperatures from -45 to 110°C (-49 to 230°F) and can be steam sterilized
The small carriers can accommodate 500 mL (16.9 oz.) or 1L (33.8 oz.) bottles. The large carriers can accommodate 2.5L (84.5 oz.) or 4L (135 oz.) bottles.
Centering ring in base assures a 2.54 cm (1") clearance between bottle and wall of carrier. Molded-in handle gives a comfortable grip.