VWR® High Quality Bibulous Paper
28511-007
- Description:VWR® high quality bibulous paper
- Size:50 sheets per block
- Cat. no.:28511-007
High quality, highly absorbent paper manufactured for minimum linting and shedding.
- Neutral paper
- Lintless drying surface
Recommended for drying slides, and small parts or components. 50 sheets per book.