Bottle Holders, White ABS, TrippNT
Proveedor: TrippNT
50080 50085 50016 50157 50189 50411 50011 50017
10010-970EA 1043.56 MXN
10010-972 11376-326 10010-970 11376-328 10010-964 11376-330 10010-968 10010-966
Bottle Holders, White ABS, TrippNT
Portadores Portadores de frascos
Store, secure and stabilize top heavy bottles with resistant bottle holders in 4 sizes. Store 500 or 1000 ml bottles securely and cleanly.
  • White chemically resistant ABS construction
  • Keeps bottles upright and secure
  • Stabilize top heavy plastic bottles
  • In stock and ready to ship
  • American made by TrippNT
  • Total replacement lifetime guarantee
Always know where to put them and when they need to be filled or replaced. This exceptionally handy holder keeps bottles secure and upright. Bright white ABS construction is chemically resistant and provides a clean, neat impression.
Holders also prevent added staining by taking the punishment instead of the bench.