Nalgene® Large Centrifuge Bio Bottles with Sealing Cap, Polypropylene Copolymer, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
3120-2024 3120-2024 3120-2006
89050-122CS 6729.89 MXN
CA89094-358 89050-122 89094-358
Nalgene® Large Centrifuge Bio Bottles with Sealing Cap, Polypropylene Copolymer, Thermo Scientific
Frascos Frascos de centrífuga
Bottles are designed for use with the Sorvall® RC12BP centrifuge and H-12000 swing-bucket rotor.
Note: Before autoclaving, place the cap on the bottle without engaging the threads. For proper performance, fill the bottle to at least 80% of the total capacity.
They are designed to process large harvests and large volume gross separations in bacterial, yeast, and tissue isolations. These graduated bottles have smooth inner walls, excellent chemical resistance, and a 50-cycle life span. Polypropylene sealing caps are autoclavable. Bottles meet USP VI requirements. They are non-cytotoxic and non-hemolytic. Bottle 89094-358 is gamma-irradiated to a Sterility Assurance Level (SAL) of 10-6 and is individually bagged.