105° Vacuum Take-Off Adapters, Chemglass
Proveedor: Chemglass
CG-1050-70 CG-1050-71 CG-1050-45 CG-1050-72 CG-1050-19 CG-1050-01 CG-1050-02 CG-1050-14
80066-054EA 1696.41 MXN
80066-058 16002-650 80066-056 80066-054 89028-244 80066-060 89028-248 89028-246
105° Vacuum Take-Off Adapters, Chemglass
Componentes para destilación Adaptadores para destilación
Adapters are bent at a 105° angle for use in vacuum distillation.
  • 105° angle bend
  • Top outer [ST] joint
  • Bottom inner [ST] joint
  • Sealed-in drip tip
  • Serrated hose connection
Adapters feature [ST] top outer and lower inner joints with sealed-in drip tip. Serrated hose connection has a 10 mm O.D. at the largest point of contact to vacuum source.