Vinyl Coat Aprons, Ansell
Proveedor: Ansell
950192 950191 950193 950194 950190
32890-884CS 7058.02 MXN
32890-884 32891-286 32890-882 32890-889 32890-888
Vinyl Coat Aprons, Ansell
Prendas desechables
SType 56-910 aprons provide excellent protection when handling acids and caustic solutions.
  • Heat-sealed binding at the neck and bottom eliminates leaks caused by exposed elastic and thread
  • Back closure consists of two plastic snaps and nylon ties, with no metal parts to cause static or corrosion
  • Full-length sleeves provide additional coverage
Made of virgin vinyl.