Stak-a-Cab™ Safety Storage Cabinets, SciMatCo
Proveedor: SciMatCo
Stak-a-Cab™
SC1560 SC2060 SC9040 SC2460 SC1460 SC9042 SC1360 SC9041 SC2260 SC2560 SC1860
14212-948EA 22194.18 MXN
14212-948 18500-058 18500-056 18500-054 14212-960 18500-062 14212-954 18500-050 18500-060 14212-958 14212-946
Stak-a-Cab™ Safety Storage Cabinets, SciMatCo
Mobiliario Armarios de almacenamiento
Choose from a variety of specialized stackable cabinets for the storage needs.
  • Cabinets are easily identified by cabinet color and (8 to 9") hazard label
  • Mini Stak-a-Cab™ cabinets offer the same level of protection in smaller units
Available as an accessory, Polypropylene isolation compartment (18500-072) fits in Stak-a-Cab™ units and isolates nitric acid or nitric acid spills from all other products stored in these cabinets.
All cabinets are constructed of 2.5cm (1") thick multi-ply plywood and finished with clear epoxy paint. Individual cabinets are designed for stacking or as stand-alone units for the laboratory bench and allow for the safe organization of compatible chemicals. All Stak-a-Cabs include polypropylene liners on the interior bottom of the cabinet for easy clean-up of accidental spills.
Certificaciones: All cabinets are supplied with suitable locking mechanisms and are constructed to meet all applicable NFPA and OSHA standards.