Nonmetallic Self-Closing Laboratory Containers, Justrite®
Proveedor: Justrite
12765 12161 12260
56610-192EA 5463.56 MXN
56610-192 56610-196 56610-190
Nonmetallic Self-Closing Laboratory Containers, Justrite®
Contenedores
Translucent, high-density polyethylene containers are specifically designed to resist acids and caustics.
  • Specially designed to resist acids and caustics
  • Translucent, high-density polyethylene containers are marked in liters, quarts, and gallons for accurate filling and dispensing
  • Spring-closing cap provides tight seal
Marked in liters, quarts, and gallons to permit accurate filling and dispensing. Spring-closing cap provides a tight seal. Available with stainless steel or plated steel fittings.