Waste Disposal Cans, HDPE, Eagle Manufacturing
1517 1525 1523 1519
56609-915EA 6364.41 MXN
56609-915 56618-514 56618-516 56609-917
Waste Disposal Cans, HDPE, Eagle Manufacturing
For temporary storage of flammable liquid wastes. Impact-resistant and will not rust or corrode.
  • For temporary storage of flammable liquid wastes
  • Impact-resistant and will not rust or corrode
  • Locking spout cap opens to an 80° angle
  • Cap assembly opens automatically to relieve excessive internal vapor pressures
  • FM approved
Fittings and flame arrester are stainless steel for superior performance
FM approved.
Spout cap opens to an 80° angle and can be locked open for convenience. Cap assembly opens automatically to relieve excessive internal vapor pressures. Choose red for disposal of flammable wastes or white for disposal of corrosive wastes.