Illuminating Coddington Magnifier, 10x, Bausch & Lomb®
Proveedor: Bausch & Lomb
813434
36933-002EA 975.1 MXN
36933-002
Illuminating Coddington Magnifier, 10x, Bausch & Lomb®
Lupas de laboratorio
With built-in light source
Direct illumination on object eliminates disturbing highlights and shadows. Requires two AA batteries (not included).