Type I Safety Cans, Galvanized Steel, Eagle Manufacturing
Proveedor: Eagle Mfg
UI20S UI50S UI20FS UI4S UI10FS UI10S UI50SB UI50SY UI50FS
56609-561EA 1720.58 MXN
56609-528 56609-517 56609-506 56618-598 56609-470 56618-596 56609-561 56609-550 56609-572
Type I Safety Cans, Galvanized Steel, Eagle Manufacturing
Contenedores
For carrying and dispensing flammable liquids safely.
Spout cap is controlled by a strong, nonrusting spring that holds the cap tight to prevent leakage or evaporation, but permits excessive gas pressure to escape and automatically closes cap when pressure is relieved. Pressing the open device allows liquid to be poured from the “no spill” pouring lip.
Made of heavy-gauge galvanized steel, finished in red baked-on powder coating. Brass flame arrester in spout absorbs and dissipates heat, keeping vapor temperature below flash point. Brass pour spout. Available in five standard Type I safety can sizes. Three sizes are available with 22.8cm (9") long HDPE detachable funnel. The 18.9L (5gal.) size is available in blue (for kerosene storage) or yellow finish (for diesel storage).
Certificaciones: UL listed. FM approved. Complies with OSHA 29 CFR 1910.106, 1925.152 and NFPA Code 30.