OptiVISOR® Binocular Headband Magnifier
Proveedor: Excelta
OptiVISOR®
DA-7 BLUE
36968-007EA 2852.02 MXN
36968-007
OptiVISOR® Binocular Headband Magnifier
Lupas de laboratorio
For close or precision work requiring visual accuracy
Continuous headband with dial adjustment for comfortable wear. Visor and lens pivot up and out of the way for normal vision when not in use. Fits over regular glasses. Magnifies with 15.2cm (6") focal length.