NovaCel™ Wipes and Sponges, Micronova
Proveedor: Micronova
NovaCel™
WP4-99
68400-131CS 33637.5 MXN
68400-131
NovaCel™ Wipes and Sponges, Micronova
Toallitas Esponjas
White PVA wipes and sponges are ideal for polishing and general wiping applications.
  • Ultraclean and chemical-resistant
  • Autoclavable
Note: Wipe 68400-850 is supplied gamma irradiated.
Firm to the touch, but soft and pliant once wetted to protect delicate surfaces and equipment.