VWR® Talon® Open Rings
Proveedor: VWR
VWR® Talon®
80862635 80862637 80862636
28641-014EA 1392.47 MXN
28641-014 28641-012 28641-010
VWR® Talon® Open Rings
Anillos de retorta
Lightweight rings mount funnels, boiling flasks, and other irregular-shaped objects to lab frames.
  • Aluminum construction
  • Lightweight
  • Easy passing of items through support area
Open ring section allows items to pass in and out of support area easily, reducing risk of breakage. Mount to rods and frames with connectors such as VWR® Talon® regular or heavy-duty holders (see 21572-501 and -566). Extension arm length: 57 mm (21/4").