Pocket Colorimeter II Test Kits, Hach
Proveedor: Hach
2506025 2668000 1406499 2506025 2106069 1407099 2105569 2508025 2527025 2105528 2106169 2105569 2802246 2503025 2668000 1406499 2508025 2507025 2242000 2105628 2770900 2430000 2770900 2105669 2106169 2242000 2503025 2105669 2502025 2802246 2507025 2504025 2106069 2430000 2105628 2527025 2502025 2105528 1407099
97009-518EA 196.21 MXN
CA97009-504 97009-518 97009-454 97009-498 CA97009-464 CA2668000 97009-530 CA97009-520 97009-456 CA97009-468 97009-436 97009-512 CA97009-488 97009-438 97009-514 CA97009-500 CA21055-28 CA97009-490 CA97009-494 CA97009-470 CA97009-514 CA97009-438 97009-520 CA97009-454 97009-464 CA97009-498 97009-500 CA97009-474 97009-488 CA97009-512 97009-502 CA97009-436 97009-468 97009-504 CA97009-456 97009-490 97009-470 97009-494 97009-474
Pocket Colorimeter II Test Kits, Hach
Kits para test medioambiente
This colorimeter is a portable, rugged, and convenient instrument, ideally suited for in-the-field analysis
Tough, impact-resistant shell is also waterproof and floats. Four buttons allow easy operation. Accuracy is comparable to expensive lab instruments. All units are factory-programmed; no manual calibration is ever required. All units come with enough reagent sets for 100 tests.