SP Bel-Art Acid and Solvent Bottle Carriers
Proveedor: Bel-Art Products, a part of SP
VWR® SP Bel-Art
F16957-0000 F16959-0000 F16960-0000 F16958-0000
56608-811CS 8897.57 MXN
56608-811 56608-900 56608-886 56608-864
SP Bel-Art Acid and Solvent Bottle Carriers
Portadores Portadores de frascos
Polyethylene carriers with epoxy coated, heavy-gauge steel wire bails have wide ribs in both the sides and on the bottom to support and cushion bottles.
  • Cone-style lid has 16mm (5/8") opening in the top for insertion of tubing, simplifies the transfer of liquids without pouring
  • Truncated style lid permits pouring without the removal of the lid
  • Designed to hold standard US reagent bottles
These acid and solvent bottle carriers are made of LDPE.