PYREX® 75° Angle Three-Way Connecting Adapter, Standard Taper Joints
Proveedor: Corning
PYREX®
9000-14 9000-19 9000-24
32645-356CS 2700 MXN
32645-356 62949-961 89091-662
PYREX® 75° Angle Three-Way Connecting Adapter, Standard Taper Joints
Adaptadores Adaptadores con juntas NS
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These angle connecting adapters have a sidearm at an approximately 75° angle from the lower joint. The lower end and sidearm are equipped with inner 24/40 standard taper joints, and the upper end with an outer 24/40 standard taper joint.