SP Bel-Art Instrument Trays With Covers, Polypropylene, Bel-Art Products, a part of SP
Proveedor: Bel-Art
F16188-0000 F16191-0000 F16189-0000
62663-266EA 4029.01 MXN
62663-266 62663-222 62663-186
SP Bel-Art Instrument Trays With Covers, Polypropylene, Bel-Art Products, a part of SP
Bandejas/Platos/Ollas Bandejas/Placas de instrumental
Trays are steam autoclavable at 121°C (250°F).
  • Easy to clean
  • Autoclavable
  • Polypropylene construction
  • Smooth, rounded corners
They have smooth, rounded corners and can be easily cleaned and sterilized. Trays taper slightly outward toward top. Each tray comes with inset cover.