VWR® Critical Cover® AlphaGuard® Lab Coats
Proveedor: VWR
VWR® Critical Cover® AlphaGuard®
LC-32632-3
10845-016CS 3562.24 MXN
10845-016
VWR® Critical Cover® AlphaGuard® Lab Coats
Prendas desechables
Premium lab coats feature a tapered collar, knit cuffs, inset sleeves, and a snap front closure.
  • Available with or without pockets
  • Excellent breathability
  • Full barrier protection
Soft, comfortable material provides excellent breathability and full barrier protection. Antistat treatment reduces static charge buildup. Lab coats are available pocketless, or with one breast and two hip pockets. Available in white.