VWR® High and Low Form Porcelain Crucibles
Proveedor: VWR
VWR®
J-124 J-500 J-138 J-141 J-118 J-139 J-134 J-136 J-117 J-122 J-119 J-135 J-502 J-137 J-501 J-123 J-120 J-121
89038-022CS 2786.97 MXN
89038-022 89038-000 89038-030 89040-208 414004-233 414004-232 414004-231 89038-028 89038-026 89038-002 89038-024 89038-034 89037-992 89037-994 89037-996 89037-988 89037-998 89037-990
VWR® High and Low Form Porcelain Crucibles
Crisoles
Chemical-porcelain crucibles are glazed inside and out, except the outside bottom and rim.
  • Glazed interior and exterior
  • Crucible covers are completely glazed, except for the rim
  • For use in temperatures up to 1150 °C (2102 °F)