Adapters with PTFE Plug, Ace Glass Incorporated
Proveedor: Ace Glass
5202-92 9080-112 5202-12 5202-110 5202-112 9080-18 9080-12 5202-114 9080-118
89051-398EA 2956.33 MXN
89082-708 89064-856 89082-770 89064-854 89082-710 89082-764 89082-736 89051-398 71001-508
Adapters with PTFE Plug, Ace Glass Incorporated
Adaptadores Conectores de tubos con juntas NS
Adapters feature either a 90° angled or straight hose connection, with ground [ST] inner joint at bottom and solid PTFE stopcock plug.
  • 90° angled or straight hose connection
  • Bottom ground [ST] inner joint
  • Solid PTFE stopcock plug
  • Manufactured from borosilicate glass
Featuring a 1:5 solid PTFE 2mm bore stopcock plug.
Información para pedidos: For replacement stopcock plug, see 89062-770 series.