KIMAX® Crystallizing Dishes, Kimble Chase
Proveedor: DWK Life Sciences
KIMAX®
23000-17090 23000-10050 23000-7050 23000-6035 23000-8040 23000-19100 23000-5035 23000-9050 23000-15075 23000-12565
89001-752CS 6989.58 MXN
89001-762 89001-752 25410-046 89001-760 25410-024 89001-758 89001-766 89001-756 89001-764 89001-754
KIMAX® Crystallizing Dishes, Kimble Chase
Bandejas/Platos/Ollas Cápsulas de evaporación
Dishes have flat bottoms and straight sides
The top of each dish is reinforced and fire polished to reduce chipping.