Aluminum Dishes with Cover, DUAL MFG CO
Proveedor: Dual Manufacturing
AD-7032 AD-5123 AD-9051 AD-6344 AD-7619
25420-028PK 1353.34 MXN
25420-028 25420-040 25420-061 25420-083 25420-072
Aluminum Dishes with Cover, DUAL MFG CO
Bandejas/Platos/Ollas Cápsulas de evaporación
Straight sides and flat bottom
Covers are interchangeable on different dishes and will fit either top or bottom of the same dish.