Synthware Vacuum Take-Off Adapters, Short Stem, Kemtech America
Proveedor: Kemtech America
Synthware
A622942 A621420 A621922 A622440
60000-638EA 911.71 MXN
60000-638 60000-634 60000-636 60000-640
Synthware Vacuum Take-Off Adapters, Short Stem, Kemtech America
Adaptadores Conectores de tubos con juntas NS
These vacuum take-off adapters have a short stem.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
  • With hose connection and drip tip
Featuring a hose connection and drip tip. Hose connection has an 8 mm ext. Ø. The top joint is outer [ST] and the bottom joint is inner [ST].