VWR® Fine-Pore Cellulose Sponges
Proveedor: VWR
VWR®
#6FP 44 66 #85FP
58540-047CT 12251.69 MXN
58540-047 58540-069 58542-360 58542-246
VWR® Fine-Pore Cellulose Sponges
Toallitas Esponjas
These fine-pore natural color cellulose sponges hold about 25 times their own weight in water.
  • Durable
  • Convenient for quick laboratory wipe-up
  • Resist tearing and abrasion
  • Absorb liquid more evenly than large pore sponges
Available in compressed (approximately ¹/₈" thickness before expansion) or uncompressed format.