VWR® PCR Workstation and Storage Box
2720-229-000 3040-500-000
24270-206CS 5773.67 MXN
24270-206 16466-026
VWR® PCR Workstation and Storage Box
The polypropylene workstation is perfect for preparing samples for cycling or working with completed procedures.
  • Includes a clear lid
  • Autoclavable
  • Freezable to -90°C (-130°F)
Workstation features a removable 96-well prep rack that holds a plate, 0.2 mL tubes, or 0.2 mL tube strips. Additional wells on the workstation accommodate 0.5 mL, 1.5 mL, and 2 mL tubes.
Información para pedidos: Pack of five workstations includes one each of red, orange, yellow, green, and blue.