Stainless Steel Metric Weight Sets, Class F, Troemner
Proveedor: Troemner
30390418 30390411 30390406 30390400
12765-495EA 22273.2 MXN
12765-490 12765-480 12765-495 12765-485
Stainless Steel Metric Weight Sets, Class F, Troemner
Pesas Pesas de calibración
Used primarily to test commercial weighing devices
Sets are supplied in a durable polypropylene case with padded handle and fitted foam interior.
Precision-crafted stainless steel weights are calibrated according to NIST specifications for Class F. NIST Class F is defined by NIST Handbook 105-1, which specifies weights used by government officials to verify commercial weighing devices. Class F is also acceptable for use in verifying scales that have a 0.1% accuracy or lower.