VWR® Basic SPP Bouffant Caps
Proveedor: VWR
VWR®
89522-670 76003-494 89522-672 89131-620 89107-768 89107-770 89107-772 89186-064 89131-622 89107-774
89522-672CS 860.38 MXN
89107-768 89522-672 89186-064 89522-670 89107-770 76003-494 89131-622 89107-772 89131-620 89107-774
VWR® Basic SPP Bouffant Caps
Prendas desechables
VWR® Basic Protection SPP Bouffant Caps are manufactured from a spunbonded polypropylene (SPP) fabric that is strong, lightweight, and breathable
  • Economical first-line barrier
  • Non-linting
Bouffant caps are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
They protect against dirt, grime, and certain dry particulates in nonhazardous environments. These general-purpose garments are ideal for less critical areas or pre-gowning entry rooms.