VWR® Weighing Funnels
Proveedor: VWR
VWR®
30287-029 329-9304 30287-040 329-9302
30287-029EA 1363.5 MXN
30287-029 CA30287-040 CA30287-029 30287-040
VWR® Weighing Funnels
Navecillas Navecillas de pesaje
Combination weighing vessel/funnel for speedy, precise weighing and transfer of samples without loss.
  • Thin-walled design
  • Glass fabrication
  • Flat base provides stable support
Made of thin-walled glass. Flattened base provides stable rest on balance pan.