Order Entry
Antistatic Weighing Canoes, Dyn-A-Med Products
Antistatic Weighing Canoes, Dyn-A-Med Products
undefined
Antistatic Weighing Canoes, Dyn-A-Med Products
  •  
    Material
    Size
    Interior Dimensions
    Color
    Supplier No.
  • 13451-986
    80051
    Polystyrene
    Small
    3.5W x 4.8L x 1.3D cm (13/8 x 17/8 x 1/2")
    Natural
    80051
    Case (250UN)
    Case (250UN) - $572.93
    $572.93
    Case (250UN)
       $572.93
    Specifications
    Environmentally Preferable
  • 13451-989
    80056
    Polystyrene
    Medium
    7.3W x 12.1L x 3.2D cm (2 7/8 x 4 3/4 x 1 1/4")
    Natural
    80056
    Paquete de 250
    Paquete de 250 - $1,544.64
    $1,544.64
    Case de 6 (250 X 250UN)
    Case de 6 (250 X 250UN) - $1,683.66
    $1,683.66
    Paquete de 250
       $1,544.64
    Case de 6 (250 X 250UN)
       $1,683.66
    Specifications
    Environmentally Preferable
  • 470202-542
    80056
    Polystyrene
    Medium
    7.3W x 12.1L x 3.2D cm (2 7/8 x 4 3/4 x 1 1/4")
    Natural
    80056
    Paquete de 250
    Paquete de 250 - $1,544.64
    $1,544.64
    Case de 6 (250 X 250UN)
    Case de 6 (250 X 250UN) - $1,683.66
    $1,683.66
    Paquete de 250
       $1,544.64
    Case de 6 (250 X 250UN)
       $1,683.66