VWR® Aluminum Micro Weighing Dishes
Proveedor: VWR
VWR®
MAWD-3500 MAWD-1120 MAWD-1201 MAWD-0400 MAWD-0350 MAWD-1100 MAWD-1200
12577-064PK 2590.8 MXN
12577-064 12577-066 12577-060 12577-070 12577-062 12577-072 12577-068
VWR® Aluminum Micro Weighing Dishes
Navecillas Navecillas de pesaje
Ideal for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use.
  • Easy to use
  • Compatible with most commercially available analyzers and microbalances
  • For use in a variety of applications
Our micro weighing boats are compatible with most commercially available analyzers and microbalances. Packaged in easy-to-use plastic containers for storage and dispensing.