Nalgene® Laboratory Notebooks, Thermo Scientific
Catalog # 51280-108
Supplier: Thermo Fisher Scientific
Specifications
- Cover Color:Forest green
- Ruling:Grid, 0.25"
- Length:281 mm (11")
- Cat. no.:51280-108
- Width:216 mm (8.5")
- Supplier No.:6301-1000
Specifications
About this item
Notebooks feature sewn-in paper pages with either horizontal ruled lines or grid
Pages have spaces for dates, signatures of workers and witnesses, and other necessary information for securing patent protection. Polyethylene cover is waterproof. Notebooks have 100 numbered pages.