VWR® Signature™ Shoe Covers made with DuPont™ Tyvek® IsoClean® Material
Proveedor: VWR
VWR® Signature™
IC451SWHLG0100VD IC451SWHXL0100VD IC451SWHMD0100VD
89128-794CS 4225.17 MXN
89128-792 89128-794 89128-790
VWR® Signature™ Shoe Covers made with DuPont™ Tyvek® IsoClean® Material
Prendas desechables
Shoe covers are manufactured using a unique flash spinning process that creates an excellent barrier against nonhazardous liquids and microorganisms as well as dry particles down to 0.5µm.
  • Excellent barrier against nonhazardous liquids and microorganisms, dry particles down to 0.5µm
  • For cleanroom use, chemically and biologically inert, tear resistant, and low linting
  • Single use, serged seams and elastic tops for a secure fit
  • Gripper™ soles provide enhanced skid-resistance and durability
Designed for cleanroom use, garments are chemically and biologically inert, tear-resistant, and low-linting. Single use shoe covers feature serged seams and elastic tops for a secure fit. Also equipped with flat, non-skid PVC soles.
Información para pedidos: Bulk packed.