5000 Series Disposable Respirators, 3M™
Proveedor: 3M
7000126024 7000052080 7000052081 7000126027 7000126025 7000052081 7000052079 7000051847 7000126025 7000051848 7000126026 7000126024 7000052080 7000126027 7000051847 7000126026 7000051848 7000052079
56222-409CS 10184.33 MXN
CA56222-408 CA56222-409 56222-408 56222-409 CA56222-422 CA56222-420 CA56222-464 56222-422 CA56222-410 CA56222-421 56222-460 56222-464 56222-420 CA56222-462 56222-421 56222-410 56222-462 CA56222-460
5000 Series Disposable Respirators, 3M™
Respiradores Mascarillas desechables Mascarillas para partículas
5000 series dual cartridge respirators are designed for maintenance-free, economical worker protection. Preassembled respirators are lightweight and well-balanced for high comfort. Unique cradle suspension holds respirator in place with minimal pressure to the back of the neck or bridge of the nose. Low-profile facepiece/cartridge provides a wider field of vision and greater comfort while wearing glasses or goggles. Color-coded by size. Resealable storage bag keeps respirator clean between uses.
  • Low-profile facepiece/cartridge for wider field of vision, greater comfort while wearing glasses or goggles
  • Color coded by size
  • Respirators suitable for gas and vapor protection come with replaceable prefilters
  • NIOSH approved
Respirators suitable for gas and vapor protection come with replaceable prefilters
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Respirator with P95 assembly comes with one pair of P95 prefilters and one pair of filter retainers.
Certificaciones: NIOSH approved.