VWR® CryoPro® Fiberboard Storage Boxes and Dividers
Proveedor: Avantor
VWR® CryoPro®
PK-A3-64 04A3-49 PK-A3-81 PK-A1-VWR-03 04A2-VWR-03 04A3-25 04A1-VWR-02M 04A2-VWR-02D 04A1-04-GEN PK-A3-25 PK-A3-25M 04A1-02M-GEN 04A2-VWR-02 PK-A3-49 PK-A3-144 04A3-100 PK-A1-VWR-02 04A1-VWR0281 04A3-64 PK-A3-16 04A1-VWR-02 PK-A3-36 04A3-81 PK-A3-100 04A3-16
82021-114PK 1218.16 MXN
82007-146 82007-144 82021-114 82021-116 82007-148 97014-322 82007-152 82007-150 89214-738 82021-122 82007-156 82021-124 89214-736 82007-154 82021-126 82007-158 82021-128 82021-120 82007-160 89214-740 82021-118 82007-142 89214-744 82007-162 89214-742
VWR® CryoPro® Fiberboard Storage Boxes and Dividers
Cajas Criocajas
Boxes are constructed of durable fiberboard with a protective, moisture-repellant coating.
  • Available with or without slots or dividers
Liquid nitrogen (LN2) freezer boxes feature slots for safe LN2 drainage and reduced LN2 consumption. Mechanical freezer boxes do not feature slots and cannot be used with LN2. Boxes with dividers feature a lid printed with a numbered grid for superior inventory control. Boxes without dividers can accomodate dividers (sold separately) for 16 to 100 cells. Fiberboard dividers can fit in any 13.2Wx13.2Lcm (51/5x51/5" ) LN2 cryogenic or mechanical freezer box.
Información para pedidos: Box dimensions include lid.