KIMAX® Liebig Condenser, Kimble Chase
Proveedor: DWK Life Sciences
KIMAX®
18140-400
89001-656CS 825.55 MXN
89001-656
KIMAX® Liebig Condenser, Kimble Chase
Condensadores
KIMAX® 400mm (153/4") full-length, one-piece sealed unit with a [ST] 24/40 outer joint at the inlet and a [ST] 24/40 inner joint at the outlet
The inlet end is sloped to avoid trapping condensate. Drip tip is provided at the lower end. Hose connections accept 7.9mm (5/16") I.D. flexible tubing. Appropriate for ASTM D849.