National™ Autosampler Vial Racks, Thermo Scientific
Proveedor: Thermo Scientific
C4012-25 C4011-25A
66030-198CS 3386.06 MXN
76212-314 66030-198
National™ Autosampler Vial Racks, Thermo Scientific
Gradillas Gradillas de viales para cromatografía
Stackable storage racks for 2 ml vials.
  • Polypropylene vial racks are resistant to most solvents
  • Efficiently hold and organize vials while using them or for storage
Racks feature alphanumeric indexing for easier vial identification. Save space by stacking the 50 or 100 plastic vial racks on top of each other.