DuPont™ Tyvek® 400 Coveralls with Respirator Hood and Attached Skid Resistant Boots, Comfort Fit Design
Proveedor: DuPont
Tyvek®
TY122SWH2X002500 TY122SWH3X002500 TY122SWH2X002500 TY122SWH7X002500 TY122SWHXL0025NF TY122SWH5X0025NF TY122SWHLG0025VP TY122SWHXL002500 TY122SWH7X0025VP TY122SWHMD002500 TY122SWH4X0025NF TY122SWHMD002500 TY122SWHXL0025VP TY122SWH2X0025NF TY122SWHXL0006G1 TY122SWH5X002500 TY122SWH4X002500 TY122SWH6X0025NF TY122SWH3X0006G1 TY122SWHMD0025NF TY122SWH4X0025VP TY122SWH3X0025VP TY122SWH2X0006G1 TY122SWH3X002500 TY122SWHLG002500 TY122SWH4X002500 TY122SWHLG0006G1 TY122SWHLG002500 TY122SWH5X0025VP TY122SWH7X0025NF TY122SWHMD0006G1 TY122SWH3X0025NF TY122SWHMD0025VP TY122SWHLG0025NF TY122SWH6X0025VP TY122SWH2X0025VP TY122SWH4X0006G1
12000-570CS 8048.42 MXN
76188-218 76188-216 76188-214 76187-750 76188-212 76188-210 76187-752 76187-926 12000-570 12000-572 76187-928 12000-574 12000-576 12000-578 CA12000-568 76188-208 76188-206 76188-204 76188-202 76188-224 76188-200 76188-222 76187-962 76188-220 76187-964 76187-966 76187-922 76187-746 76187-924 76187-968 76187-748 CA12000-570 10016-698 CA12000-574 12000-568 CA12000-576 CA12000-578
DuPont™ Tyvek® 400 Coveralls with Respirator Hood and Attached Skid Resistant Boots, Comfort Fit Design
Prendas desechables
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Comfort fit design
  • Respirator fit hood
  • Zipper front
  • Elastic wrists
  • Attached skid-resistant boots
  • Elastic waist
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Comfort fit design is based on extensive wearer input, to provide our most comfortable garment design that enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes longer zipper front for easier donning and doffing and an elastic waist to better position the garment. Attached respirator-fit hood features elastic around face opening, which is designed to cover neck and chin and fit around respirator face mask. Longer zipper extends to chin for complete coverage of neck area. Elastic openings at wrists allow for a tighter fit. Attached Tyvek® FC boots have special coating to provide added skid resistance.
Certificaciones: Thickness meets ASTM D1777.