VWR® Talon® Cylinder Wall Brackets
Proveedor: VWR
VWR® Talon®
30400025 80863370 80863378 30400029
60141-124EA 1474.29 MXN
60141-126 97035-536 97035-534 60141-124
VWR® Talon® Cylinder Wall Brackets
Colgadores de pared
Cylinder wall-mount brackets are constructed of cast aluminum and contoured to allow cylinders to fit firmly along the support edge.
  • Cast aluminum construction
  • Nickel-plated, non-slip spring catch and buckle
  • Easily adjustable
Recessed screw holes at each side of the bracket allow for easy wall mounting. Features a 2.5 cm (1") wide, 137.2 cm (54") long nylon strap with nickel-plated, non-slip spring catch and buckle for fast, easy adjustment. For cylinders from 10.2 – 35.6 cm (4 – 14") in diameter. Available with or without a “Secure Cylinder” safety message strap.