VWR® Four-Way Microtube Racks
Proveedor: Avantor
VWR®
HS29022AV HS29022EV HS29022FV HS29022DV HS29022CV HS29022GV
82024-458CS 3608.22 MXN
82024-458 82010-752 82024-462 82024-460 82024-464 82010-750
VWR® Four-Way Microtube Racks
Gradillas Tube Racks
These heavy-duty, polypropylene racks link together to meet custom rack needs.
  • Autoclavable
  • Polypropylene fabrication
Racks feature an interlocking system of tabs and slots for easy connection and sturdy fit. Each rack has four sides that hold either four 50 mL conical tubes, twelve 15 mL conical tubes, thirty-two 1.5 mL microtubes, or thirty-two 0.5 mL microtubes.
Información para pedidos: Assorted rack pack 82010-750 includes one each in blue, green, pink, yellow, and orange.