DuPont™ Tyvek® 400 Coveralls with Respirator Hood and Elastic Wrists and Ankles, Comfort Fit Design
Proveedor: DuPont
Tyvek®
TY127SWH3X002500 TY127SWHMD002500 TY127SWH7X0025VP TY127SWH5X0025NF TY127SWHLG0006G1 TY127SWHXL0006G1 TY127SWH6X002500 TY127SWH4X002500 TY127SWH3X002500 TY127SWH2X0025NF TY127SWHLG002500 TY127SWHMD0025VP TY127SWH2X002500 TY127SWH2X0025VP TY127SWH4X0025NF TY127SWHMD0025NF TY127SWH5X002500 TY127SWH7X002500 TY127SWH6X0025NF TY127SWHXL002500 TY127SWHMD0006G1 TY127SWHLG0025NF TY127SWHXL0025VP TY127SWHLG002500 TY127SWH3X0025VP TY127SWH5X002500 TY127SWH4X002500 TY127SWH3X0025NF TY127SWHLG0025VP TY127SWHXL0025NF TY127SWH2X002500 TY127SWH2X0006G1 TY127SWH4X0006G1 TY127SWH7X0025NF TY127SWH3X0006G1 TY127SWH4X0025VP TY127SWH6X0025VP TY127SWHMD002500 TY127SWH5X0025VP TY127SWHXL002500
12000-580CS 6688.93 MXN
76188-258 76188-256 76188-278 76188-276 76188-254 76188-274 76188-252 76188-272 76188-250 76188-270 13452-890 12000-590 CA12000-580 CA12000-582 CA13452-890 CA12000-584 CA13452-892 CA12000-586 CA12000-588 76188-248 76188-246 76188-268 10016-700 76188-244 76188-288 76188-266 76188-264 76188-286 76188-242 76188-284 76188-262 12000-580 76188-260 76188-282 12000-582 76188-280 CA12000-590 12000-584 12000-586 12000-588
DuPont™ Tyvek® 400 Coveralls with Respirator Hood and Elastic Wrists and Ankles, Comfort Fit Design
Prendas desechables
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Comfort fit design
  • Respirator fit hood
  • Zipper front
  • Elastic wrists and ankles
  • Elastic waist
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Comfort fit design is based on extensive wearer input, to provide our most comfortable garment design that enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes longer zipper front for easier donning and doffing and an elastic waist to better position the garment. Attached respirator-fit hood features elastic around face opening, which is designed to cover neck and chin and fit around respirator face mask. Longer zipper extends to chin for complete coverage of neck area. Elastic openings at wrists and ankles allow for a tighter fit.
Certificaciones: Thickness meets ASTM D1777.