SP Bel-Art Serum Vial Racks, Polypropylene, Bel-Art Products, a part of SP
Proveedor: Bel-Art
SCIENCEWARE®
F18513-1640 F18513-2040 F18513-0072
60985-476EA 1046.89 MXN
60985-476 60985-498 60985-454
SP Bel-Art Serum Vial Racks, Polypropylene, Bel-Art Products, a part of SP
Gradillas Gradillas de viales para cromatografía
Just the right size for serum vials, RIA vials and T3/T4 reaction vials.
  • Polypropylene rack fabrication
  • Corrosion-resistant
  • Long effective lifepsan
  • Autoclavable
Long-lasting and corrosion-resistant, these polypropylene racks fit most water baths for elevated temperature incubations. Top shelf height is 41 mm (158"). End plates have handles.