Nalgene® Tubing Connectors, Polypropylene, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
6152-0500 6152-0250 6152-0312 6151-0500 6151-0250 6152-0375 6151-0187 6151-0375 6151-0125 6152-0187 6151-0312 6152-0125
62860-063CS 6269.16 MXN
62850-128 62860-063 62850-106 62860-041 62860-085 62860-020 62850-081 62850-060 62860-100 62860-121 62850-048 62850-026
Nalgene® Tubing Connectors, Polypropylene, Thermo Scientific
Racores y conectores
Durable polypropylene connectors are resistant to gases, solvents, and acids
Available in T- and Y-type connectors. Autoclavable.