DuPont™ Tyvek® 400 Coveralls with Open Wrists and Ankles, Zipper Front Closure, Comfort Fit Design
Proveedor: DuPont
Tyvek®
TY120SWH6X0025NF TY120SWH7X0025VP TY120SWHMD0006G1 TY120SWHMD002500 TY120SWH2X002500 TY120SWH4X002500 TY120SWHLG0025NF TY120SWH4X0006G1 TY120SWH5X002500 TY120SWH6X002500 TY120SWH5X0025NF TY120SWH3X0006G1 TY120SWH4X0025VP TY120SWH2X0025NF TY120SWHLG002500 TY120SWHMD0025NF TY120SWHXL0025NF TY120SWHMD002500 TY120SWH6X0025VP TY120SWH7X002500 TY120SWHXL0006G1 TY120SWH2X0006G1 TY120SWH3X002500 TY120SWHLG0025VP TY120SWH2X002500 TY120SWHLG002500 TY120SWH4X002500 TY120SWHXL0025VP TY120SWH7X0025NF TY120SWH3X0025VP TY120SWH3X002500 TY120SWHXL002500 TY120SWHXL002500 TY120SWHLG0006G1 TY120SWH4X0025NF TY120SWH5X0025VP TY120SWH2X0025VP TY120SWHMD0025VP TY120SWH3X0025NF
76188-158 76188-178 76188-156 76188-176 76188-154 76188-152 76188-174 76188-196 76188-172 76188-194 76188-150 76188-192 CA13450-542 76188-170 CA13450-544 76188-190 13450-534 CA13450-540 13450-538 13450-536 10016-906 76188-168 76188-166 76188-188 76188-186 76188-164 76188-162 10016-902 76188-184 76188-182 10016-904 76188-160 13450-542 76188-180 13450-540 13450-544 CA13450-538 CA13450-534 CA13450-536
DuPont™ Tyvek® 400 Coveralls with Open Wrists and Ankles, Zipper Front Closure, Comfort Fit Design
Prendas desechables
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Comfort fit design
  • Laydown collar
  • Zipper front
  • Open wrists and ankles
  • Elastic waist
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Comfort fit design is based on extensive wearer input, to provide our most comfortable garment design that enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes longer zipper front for easier donning and doffing and an elastic waist to better position the garment. Coveralls feature a laydown collar, zipper front, and open wrists and ankles.
Certificaciones: Thickness meets ASTM D1777.