WHEATON® Aluminum Seals (Unlined), DWK Life Sciences
Proveedor: DWK Life Sciences
224178-05 224189 W224205 224177-05 W224207 224177-06 224176-01 224193-06
10796-546CS 2160.7 MXN
10796-544 10796-174 10796-628 10796-540 10796-630 10796-542 10796-538 10796-546
WHEATON® Aluminum Seals (Unlined), DWK Life Sciences
Tapas Tapones para viales de cromatografía
Aluminum seals (unlined) are available in a wide range of styles and sizes.
  • Open top seals expose stopper for sample retrieval with a syringe
  • Tear-off seal removes completely allowing content to pour from bottle
  • Solid top seals are excellent for long-term storage of samples
  • Flip cap seals provide tamper evidence
Color selection includes natural, red, blue, and green.
Envase: Shelf packs of 100.