DicksonWare™ Software, Dickson
Proveedor: Dickson
DicksonWare™
A015 A026 A016 A025
14905-016EA 7498.51 MXN
12444-034 14905-016 13450-310 14905-014
DicksonWare™ Software, Dickson
Software Software de laboratorio
Software allows for simple viewing and zooming of logged data.
The software is also available in secure, 21CFR11-compliant version. This version provides password protection, data encryption, and an audit trail of user activity. Secure software labels all charts with the user ID and serial number of data logger used.
Software features user selectable sample intervals, delayed logger start times, and temperature displays in Celsius or Fahrenheit. Data and graphs can be downloaded quickly, exported effortlessly, and viewed in tabular or graphical formats. Allows for real-time monitoring. Microsoft Windows® compatible. Software includes USB or serial download cable.